Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024504.2 | NP_001019675.2 | 7 | 2865 | ||
NM_001042649.1 | NP_001036114.1 | 6 | 2782 | |||
NM_001042650.1 | NP_001036115.1 | 5 | 2685 | |||
NM_001042651.1 | NP_001036116.1 | 6 | 2768 | |||
GenBank mRNA | AK040130.1 | - | 3 | 2297 | ||
AK154550.1 | - | 6 | 2788 | |||
AK155726.1 | - | 7 | 2581 | |||
AK161694.1 | - | 6 | 2474 | |||
BC030839.1 | - | 7 | 2573 | |||
BC052676.1 | - | 5 | 2687 | |||
DV648583.1 | - | 1 | 100 |