Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038695.3 | NP_001033784.2 | 10 | 1079 | ||
NM_173001.3 | NP_766589.1 | 9 | 1106 | |||
GenBank mRNA | AK082379.1 | - | 4 | 893 | ||
AK129204.1 | - | 10 | 1067 | |||
AK144825.1 | - | 9 | 1107 | |||
BC032256.1 | - | 10 | 1055 | |||
BC049876.1 | - | 3 | 149 | |||
BC059264.1 | - | 9 | 989 | 6409698 | ||
DQ323991.1 | - | 6 | 544 |