Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035233.1 | NP_001030310.1 | 1383 | |||
NM_017215.2 | NP_058911.2 | 1383 | ||||
GenBank mRNA | AF297648.1 | - | 1295 | |||
AF451299.1 | - | 755 | ||||
AF465909.1 | - | 1276 | ||||
AY044832.1 | - | 1272 | ||||
AY069978.1 | - | 574 | ||||
AY578981.1 | - | 1 | 26 | |||
JN132400.1 | - | 1458 | ||||
U15098.1 | - | 1332 | ||||
X67857.1 | - | 1370 |