Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081043.1 | NP_001074512.1 | 2, 3 | 257 | ||
GenBank mRNA | CF538443.1 | - | 2, 3 | 218 |