Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142152.1 | - | 6 | 766 | ||
AK154645.1 | - | 4 | 2367 | |||
AK163238.1 | - | 4 | 2351 | |||
BC031484.1 | - | 6 | 761 | 2654265 | ||
BC047372.1 | - | 6 | 744 | |||
BC057597.1 | - | 4 | 2329 | |||
BC089379.1 | - | 6 | 719 |