Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039521.1 | NP_001034610.1 | 18 | 3349 | ||
GenBank mRNA | AK080397.1 | - | 18 | 3312 | ||
AK165676.1 | - | 18 | 3334 | |||
AK170340.1 | - | 3 | 1667 | |||
AK172693.1 | - | 463 | ||||
BC034110.1 | - | 14 | 2864 | |||
BC055781.1 | - | 5 | 1967 |