Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276292.1 | NP_001263221.1 | 24 | 4752 | ||
NM_177327.5 | NP_796301.2 | 25 | 4914 | |||
GenBank mRNA | AK033138.1 | - | 17 | 3729 | ||
AK042570.1 | - | 1 | 1426 | |||
AK049113.1 | - | 1 | 144 | |||
AK140411.1 | - | 1 | 1891 | |||
BC051405.1 | - | 4 | 2231 |