Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081414.2 | NP_001074883.1 | 10 | 7900 | ||
NM_001143834.1 | NP_001137306.1 | 11 | 7996 |
Set Membership: |
G-protein coupled receptor |