Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem74b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160363.1 | NP_001153835.1 | 2 | 1223 | ||
GenBank mRNA | AK077378.1 | - | 2 | 1224 | ||
BC076500.1 | - | 1 | 421 |