Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053009.3 | NP_443735.2 | 11 | 4474 | ||
GenBank mRNA | AK051904.1 | - | 6 | 2847 | ||
AK088933.1 | - | 7 | 3035 | |||
AK147502.1 | - | 11 | 3787 | |||
AK147560.1 | - | 11 | 4478 | |||
AK161468.1 | - | 1 | 1716 | |||
BC057323.1 | - | 7 | 3013 | |||
BC083000.1 | - | 7 | 3013 |