Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053009.3 | NP_443735.2 | 11 | 4888 | ||
GenBank mRNA | AK088933.1 | - | 7 | 3450 | ||
AK147502.1 | - | 11 | 4201 | |||
AK147560.1 | - | 11 | 4892 | |||
AK161468.1 | - | 1 | 2130 | |||
BC057323.1 | - | 7 | 3427 | |||
BC083000.1 | - | 7 | 3427 |