Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017240.2 | NP_058936.1 | 1 | 102 | ||
XM_006251951.2 | XP_006252013.1 | 3 | 252 | |||
GenBank mRNA | X15939.1 | - | 1 | 104 |