Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081037.2 | NP_001074506.1 | 22 | 6945 | ||
NM_001242411.1 | NP_001229340.1 | 22 | 6876 |
Gene Symbol: C030027H14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021106.1 | - | 1 | 156 |