Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009742.3 | NP_033872.1 | 1 | 465 | ||
GenBank mRNA | L16462.1 | - | 1 | 465 |