Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017244.1 | NP_058940.1 | 257 | |||
GenBank mRNA | U23407.1 | - | 257 |
Gene Symbol: LOC100911902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749317.3 | XP_003749365.1 | 2 | 1003 |