Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bglap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037939.2 | NP_001033028.1 | 1 | 61 | ||
NM_007541.3 | NP_031567.1 | 1 | 61 | |||
GenBank mRNA | BY341522.1 | - | 1 | 59 | ||
BY351185.1 | - | 1 | 60 | |||
CJ326308.1 | - | 1 | 62 | |||
U11542.1 | - | 1 | 28 |