Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010057.2 | NP_034187.1 | 1 | 91 | ||
GenBank mRNA | EF535989.1 | - | 1 | 91 | ||
EF535990.1 | - | 1 | 91 |