Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081011.2 | NP_001074480.2 | 23 | 4728 | ||
GenBank mRNA | AK132220.1 | - | 9 | 2401 | ||
AK160574.1 | - | 1 | 549 | |||
BC151081.1 | - | 22 | 4346 | 9087994 | ||
BC151082.1 | - | 22 | 4346 | 9087995 | ||
BC158055.1 | - | 22 | 4344 | 9088163 | ||
BC172152.1 | - | 22 | 4344 | 9054822 |