Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081081.2 | NP_001074550.1 | 6, 7 | 1232 | ||
NM_001113383.1 | NP_001106854.1 | 6, 7 | 1232 | |||
GenBank mRNA | AK046297.1 | - | 1, 2 | 154 | ||
AK048072.1 | - | 4, 5 | 410 | |||
AK141044.1 | - | 2, 3 | 218 | |||
AK173039.1 | - | 6, 7 | 1232 |