Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081097.2 | NP_001074566.1 | 16 | 2668 | ||
GenBank mRNA | AF245444.1 | - | 8 | 1463 | ||
BC118004.1 | - | 16 | 2663 |