Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130693.3 | NP_001124165.1 | 17 | 3176 | ||
GenBank mRNA | AK131782.1 | - | 3 | 145 |