Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243712.1 | NP_001230641.1 | 26 | 5317 | ||
NM_008395.3 | NP_032421.2 | 25 | 5115 | |||
GenBank mRNA | AF037454.1 | - | 25 | 5027 | ||
AK051021.1 | - | 1 | 987 | |||
AK087794.1 | - | 1 | 1973 | |||
AK220164.1 | - | 26 | 5235 | |||
BC062934.1 | - | 25 | 5064 | 30535756 | ||
BC064678.1 | - | 24 | 5093 | 6409535 | ||
BF453521.1 | - | 1 | 313 |