Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010690.1 | NP_034820.1 | 8 | 5154 | ||
GenBank mRNA | AK029530.1 | - | 1 | 606 | ||
AK081925.1 | - | 1 | 391 | |||
AK151387.1 | - | 1 | 819 |