Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010806.1 | NP_034936.1 | 33 | 5553 | ||
GenBank mRNA | BC026648.1 | - | 3 | 831 | ||
BI991173.1 | - | 3 | 392 |