Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173506.1 | NP_001166977.1 | 4 | 4660 | ||
NM_032393.2 | NP_115769.1 | 5 | 4944 | |||
GenBank mRNA | AK147474.1 | - | 1 | 3156 |