Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003883.3 | NP_003874.2 | 15 | 1388 | ||
XM_011537697.2 | XP_011535999.1 | 14 | 1240 | |||
GenBank mRNA | AF005482.1 | - | 13 | 1404 | ||
AF039703.1 | - | 15 | 1344 | |||
BC000614.1 | - | 15 | 1377 | 3345755 | ||
U66914.1 | - | 15 | 1333 | |||
U75696.1 | - | 14 | 1371 | |||
U75697.1 | - | 15 | 1377 |