Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081327.1 | NP_001074796.1 | 2 | 1479 | ||
GenBank mRNA | AK032292.1 | - | 1 | 403 |