Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090073.1 | - | 7 | 816 | ||
AK148298.1 | - | 7 | 816 | |||
AK173247.1 | - | 8 | 1056 | |||
AK190037.1 | - | 2 | 134 | |||
S79463.1 | - | 7 | 670 |