Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206897.1 | NP_001193826.1 | 9 | 1243 | ||
NM_003888.3 | NP_003879.2 | 8 | 1146 | |||
NM_170696.2 | NP_733797.1 | 7 | 1032 | |||
NM_170697.2 | NP_733798.1 | 6 | 750 | |||
GenBank mRNA | AB015226.1 | - | 8 | 916 | ||
AB015227.1 | - | 6 | 679 | |||
AB015228.1 | - | 6 | 641 | |||
AK128709.1 | - | 8 | 1641 | |||
AK294981.1 | - | 8 | 846 | |||
AK303057.1 | - | 9 | 988 | |||
BC030589.2 | - | 7 | 790 | 4826743 | ||
BX443600.2 | - | 6 | 751 |