Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252569.1 | NP_001239498.1 | 6 | 1427 | ||
NM_009243.4 | NP_033269.1 | 5 | 1212 | |||
GenBank mRNA | AK146619.1 | - | 1145 | |||
BC011040.1 | - | 1138 | 4159448 | |||
BC037007.1 | - | 1364 | 5123904 | |||
BC047128.1 | - | 1138 | ||||
BC049970.1 | - | 1138 | 4160332 | |||
BC057982.1 | - | 1145 | 5134754 | |||
BC057984.1 | - | 1160 | 5126441 | |||
BC057989.1 | - | 1123 | 5100593 | |||
M75721.1 | - | 1143 | ||||
W82055.1 | - | 2 | 141 |