Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcpkmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033236.2 | NP_001028408.2 | 6 | 972 | ||
GenBank mRNA | AK144832.1 | - | 6 | 938 |