Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033240.4 | NP_001028412.1 | 3 | 272 | ||
NM_001177848.1 | NP_001171319.1 | 2 | 281 | |||
GenBank mRNA | AK136589.1 | - | 2 | 282 | ||
BC147597.1 | - | 2 | 255 | 9056587 | ||
BC147615.1 | - | 2 | 255 | 9056605 | ||
BM933122.1 | - | 3 | 274 |