Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081370.2 | NP_001074839.2 | 11 | 3913 | ||
NM_001113373.2 | NP_001106844.2 | 16 | 4359 | |||
GenBank mRNA | AB099695.1 | - | 16 | 3998 | ||
AK220363.1 | - | 9 | 2928 | |||
CF540072.1 | - | 2 | 302 |