Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285927.1 | NP_001272856.1 | 11 | 1362 | ||
NM_175291.4 | NP_780500.2 | 11 | 1346 | |||
GenBank mRNA | AK042052.1 | - | 11 | 1321 | ||
AK122347.1 | - | 2 | 200 | |||
AK138722.1 | - | 11 | 1348 | |||
BC150753.1 | - | 11 | 1282 | 9087664 | ||
GU797481.1 | - | 11 | 1319 |