Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081173.1 | NP_001074642.1 | 2 | 420 | ||
GenBank mRNA | AK144916.1 | - | 2 | 250 |