Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081173.1 | NP_001074642.1 | 19 | 2032 | ||
GenBank mRNA | AK122516.1 | - | 7 | 1796 | ||
BC056180.1 | - | 13 | 986 |