Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081040.1 | NP_001074509.1 | 2 | 1416 | ||
GenBank mRNA | AK080862.1 | - | 1 | 1106 | ||
BC147716.1 | - | 2 | 565 | |||
BC147722.1 | - | 2 | 565 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK346052.1 | - | 88 | Chr. - - [-] on Build |