Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102565.1 | NP_001096035.1 | 6 | 1679 | ||
GenBank mRNA | AK143105.1 | - | 8 | 1889 | ||
AK144828.1 | - | 6 | 1678 | |||
AK146582.1 | - | 1097 | ||||
BC027086.1 | - | 4 | 1171 | |||
BC050856.1 | - | 7 | 1765 | |||
BC094381.1 | - | 6 | 1613 |