Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017298.1 | NP_058994.1 | 1, 2 | 431 | ||
XM_006252586.2 | XP_006252648.1 | 1, 2 | 493 | |||
XM_006252590.2 | XP_006252652.1 | 1, 2 | 493 | |||
XM_006252594.2 | XP_006252656.1 | 1, 2 | 493 | |||
XM_006252595.2 | XP_006252657.1 | 1, 2 | 493 | |||
XM_006252597.2 | XP_006252659.1 | 1, 2 | 493 | |||
XM_008770991.1 | XP_008769213.1 | 1, 2 | 493 | |||
XM_008770992.1 | XP_008769214.1 | 1, 2 | 589 | |||
GenBank mRNA | AF370009.1 | - | 1, 2 | 50 | ||
D38101.1 | - | 1, 2 | 431 | |||
D38102.1 | - | 1, 2 | 431 | |||
M57682.1 | - | 1, 2 | 600 |