Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033245.4 | NP_001028417.1 | 1 | 178 | ||
NM_001206392.1 | NP_001193321.1 | 1 | 178 | |||
GenBank mRNA | AK149651.1 | - | 1 | 28 | ||
AK154603.1 | - | 1 | 150 | |||
AK155701.1 | - | 1 | 91 | |||
AK159857.1 | - | 1 | 28 | |||
AK159906.1 | - | 1 | 88 | |||
AK160018.1 | - | 1 | 88 | |||
AK160063.1 | - | 1 | 88 | |||
AK162637.1 | - | 1 | 121 | |||
AK170813.1 | - | 1 | 28 | |||
AK171214.1 | - | 1 | 116 | |||
AK171248.1 | - | 1 | 28 | |||
AK171580.1 | - | 1 | 128 |