Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pm20d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178079.3 | NP_835180.2 | 8, 9 | 996 | ||
GenBank mRNA | AK028883.1 | - | 8, 9 | 993 | ||
AK143953.1 | - | 8, 9 | 991 | |||
BC025830.1 | - | 5, 6 | 486 | |||
BC049922.1 | - | 1, 2 | 33 | |||
BC120725.1 | - | 8, 9 | 996 | 40129648 | ||
BC125395.1 | - | 8, 9 | 996 | 40129910 |