Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam46a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160378.1 | NP_001153850.1 | 3 | 2724 | ||
NM_001160379.1 | NP_001153851.1 | 3 | 2695 | |||
GenBank mRNA | AK032747.1 | - | 1 | 481 |