Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017309.2 | NP_059005.1 | 3 | 282 | ||
XM_006251517.2 | XP_006251579.2 | 3 | 325 | |||
XM_008770426.1 | XP_008768648.1 | 2 | 995 | |||
GenBank mRNA | BC088855.1 | - | 3 | 282 | 7317429 | |
D14425.1 | - | 3 | 250 | |||
D14568.1 | - | 3 | 138 | |||
FQ212066.1 | - | 3 | 449 | |||
L03554.1 | - | 2 | 84 |