Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033253.3 | NP_001028425.1 | 16 | 6316 | ||
NM_001159942.1 | NP_001153414.1 | 17 | 6482 | |||
GenBank mRNA | AK076288.1 | - | 1 | 1955 | ||
AK122468.1 | - | 13 | 5640 | |||
BC076628.1 | - | 16 | 4501 |