Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164225.1 | NP_001157697.1 | 6 | 3269 | ||
GenBank mRNA | BC042620.1 | - | 5 | 2978 |