Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017315.2 | NP_059011.1 | 4, 5 | 433 | ||
XM_006254602.2 | XP_006254664.1 | 6, 7 | 814 | |||
GenBank mRNA | AF080452.1 | - | 4, 5 | 433 | ||
BC078851.1 | - | 4, 5 | 433 | 7127602 |