Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081089.2 | NP_001074558.2 | 12 | 1303 | ||
GenBank mRNA | AB257502.1 | - | 11 | 1221 | ||
AK131154.1 | - | 12 | 1302 | |||
BC113165.1 | - | 12 | 1297 | 40059976 | ||
BC113776.1 | - | 12 | 1297 | 40059969 |