Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024539.1 | NP_001019710.1 | 13 | 3196 | ||
GenBank mRNA | BC059866.1 | - | 2 | 3072 |