Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045521.1 | NP_001038986.1 | 5 | 877 | ||
GenBank mRNA | AK145810.1 | - | 5 | 882 | ||
BC147400.1 | - | 5 | 873 | 9056455 | ||
BC147401.1 | - | 5 | 873 | 9056447 | ||
BY757078.1 | - | 3 | 333 |