Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC030499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287206.1 | NP_001274135.1 | 1 | 16 | ||
GenBank mRNA | BY266382.1 | - | 1 | 17 |